[2-(Methylamino)-5-nitrophenyl]methanol ≥95%

Supplier: Matrix Scientific

054273-500MG 054273-1G
101906-860EA 375.38 USD
101906-860 101906-862
[2-(Methylamino)-5-nitrophenyl]methanol ≥95%
[2-(Methylamino)-5-nitrophenyl]methanol

Hazard: Irritant

Formula: C₈H₁₀N₂O₃
MDL Number: MFCD14581654
CAS Number: 946582-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR