1-(Dimethylamino)-4-methyl-1-penten-3-one ≥95%

Supplier: Matrix Scientific

054272-500MG 054272-1G
101906-854EA 307.69 USD
101906-854 101906-856
1-(Dimethylamino)-4-methyl-1-penten-3-one ≥95%
1-(Dimethylamino)-4-methyl-1-penten-3-one

Hazard: Irritant

Formula: C₈H₁₅NO
MDL Number: MFCD09859309
CAS Number: 5782-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR