Methyl-5-bromo-2-fluoronicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-2-fluoronicotinate

054271-250MG 054271-1G 054271-5G
101906-848EA 1250.77 USD
101906-848 101906-850 101906-852
Methyl-5-bromo-2-fluoronicotinate ≥95%
Methyl-5-bromo-2-fluoronicotinate

Hazard: Irritant

Formula: C₇H₅BrFNO₂
MW: 234.0200 g/mol
MDL Number: MFCD11617316
CAS Number: 931105-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR