2-Chloro-6-isopropyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine ≥95%

Supplier: Matrix Scientific

054270-500MG 054270-1G
101906-844EA 921.54 USD
101906-844 101906-846
2-Chloro-6-isopropyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine ≥95%
2-Chloro-6-isopropyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine

Hazard: Irritant

Formula: C₁₁H₉ClF₃N₃O
MDL Number: MFCD14581653
CAS Number: 1203897-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR