2-Chloro-3-(5-methyl-1,2,4-oxadiazol-3-yl)-6-phenylpyridine ≥95%

Supplier: Matrix Scientific

054269-500MG 054269-1G
101906-840EA 921.54 USD
101906-840 101906-842
2-Chloro-3-(5-methyl-1,2,4-oxadiazol-3-yl)-6-phenylpyridine ≥95%
2-Chloro-3-(5-methyl-1,2,4-oxadiazol-3-yl)-6-phenylpyridine

Hazard: Irritant

Formula: C₁₄H₁₀ClN₃O
MDL Number: MFCD14581652
CAS Number: 1203898-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR