2-Chloro-N'-hydroxy-6-isopropyl-3-pyridinecarboximidamide ≥95%

Supplier: Matrix Scientific

054268-500MG 054268-1G
101906-836EA 556.92 USD
101906-836 101906-838
2-Chloro-N'-hydroxy-6-isopropyl-3-pyridinecarboximidamide ≥95%
2-Chloro-N'-hydroxy-6-isopropyl-3-pyridinecarboximidamide

Hazard: Irritant

Formula: C₉H₁₂ClN₃O
MW: 213.67 g/mol
MDL Number: MFCD14581651
CAS Number: 1198595-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR