2-Chloro-6-phenyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine ≥95%

Supplier: Matrix Scientific

054266-500MG 054266-1G
101906-828EA 921.54 USD
101906-828 101906-830
2-Chloro-6-phenyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine ≥95%
2-Chloro-6-phenyl-3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]pyridine

Hazard: Irritant

Formula: C₁₄H₇ClF₃N₃O
MW: 325.68 g/mol
MDL Number: MFCD14581650
CAS Number: 1198475-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR