2-Chloro-N'-hydroxy-6-phenyl-3-pyridinecarboximidamide ≥95%

Supplier: Matrix Scientific

054265-500MG 054265-1G
101906-824EA 747.69 USD
101906-824 101906-826
2-Chloro-N'-hydroxy-6-phenyl-3-pyridinecarboximidamide ≥95%
2-Chloro-N'-hydroxy-6-phenyl-3-pyridinecarboximidamide

Hazard: Irritant

Formula: C₁₂H₁₀ClN₃O
MW: 247.69 g/mol
MDL Number: MFCD14581649
CAS Number: 1198595-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR