6-Phenyl-2-piperazinonicotinonitrile ≥95%

Supplier: Matrix Scientific

054264-500MG 054264-1G
101906-820EA 747.69 USD
101906-820 101906-822
6-Phenyl-2-piperazinonicotinonitrile ≥95%
6-Phenyl-2-piperazinonicotinonitrile

Hazard: Irritant

Formula: C₁₆H₁₆N₄
MDL Number: MFCD14581648
CAS Number: 1198475-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR