2,3-Dichloro-5-(trifluoromethyl)isonicotinamide ≥95%

Supplier: Matrix Scientific

054262-500MG 054262-1G
101906-812EA 747.69 USD
101906-812 101906-814
2,3-Dichloro-5-(trifluoromethyl)isonicotinamide ≥95%
2,3-Dichloro-5-(trifluoromethyl)isonicotinamide

Hazard: Irritant

Formula: C₇H₃Cl₂F₃N₂O
MDL Number: MFCD14581646
CAS Number: 1198475-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR