1,2-Diphenyl-1-ethanone oxime ≥95%

Supplier: Matrix Scientific

054261-500MG 054261-1G 054261-5G
101906-806EA 103.08 USD
101906-806 101906-808 101906-810
1,2-Diphenyl-1-ethanone oxime ≥95%
1,2-Diphenyl-1-ethanone oxime

Hazard: Irritant

Formula: C₁₄H₁₃NO
Melting Pt: 95…97 °C
MDL Number: MFCD14581645
CAS Number: 952-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR