tert-Butyl-4-(3-cyano-6-phenyl-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
054260-500MG
054260-1G
101906-802EA
747.69
USD
101906-802
101906-804
tert-Butyl-4-(3-cyano-6-phenyl-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(3-cyano-6-phenyl-2-pyridinyl)tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₂₁H₂₄N₄O₂ MW: 364.45 g/mol |
MDL Number:
MFCD09027917 CAS Number: 946385-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...