Methyl-4-(6-amino-5-nitro-3-pyridinyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

054259-500MG 054259-1G
101906-798EA 556.92 USD
101906-798 101906-800
Methyl-4-(6-amino-5-nitro-3-pyridinyl)benzenecarboxylate ≥95%
Methyl-4-(6-amino-5-nitro-3-pyridinyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₁N₃O₄
MW: 273.25 g/mol
MDL Number: MFCD14560552
CAS Number: 1206523-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR