2,3-Dichloro-5-(trifluoromethyl)isonicotinoyl chloride ≥95%
Supplier: Matrix Scientific
054258-500MG
054258-1G
101906-794EA
747.69
USD
101906-794
101906-796
2,3-Dichloro-5-(trifluoromethyl)isonicotinoyl chloride ≥95%
2,3-Dichloro-5-(trifluoromethyl)isonicotinoyl chloride
Hazard: Irritant
Formula:
C₇HCl₃F₃NO |
MDL Number:
MFCD14581644 CAS Number: 1198475-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...