2,2,2-Trifluoro-1-(1H-pyrrol-3-yl)-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoroacetyl)-1H-pyrrole

054257-500MG 054257-1G
101906-790EA 556.92 USD
101906-790 101906-792
2,2,2-Trifluoro-1-(1H-pyrrol-3-yl)-1-ethanone ≥95%
2,2,2-Trifluoro-1-(1H-pyrrol-3-yl)-1-ethanone

Hazard: Irritant

Formula: C₆H₄F₃NO
MW: 163.1 g/mol
MDL Number: MFCD12924726
CAS Number: 130408-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR