2,5-Bis(benzyloxy)benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

054251-1G 054251-5G
101906-770EA 232.31 USD
101906-770 101906-772
2,5-Bis(benzyloxy)benzenecarbaldehyde ≥95%
2,5-Bis(benzyloxy)benzenecarbaldehyde

Hazard: Irritant

Formula: C₂₁H₁₈O₃
MDL Number: MFCD02380367
CAS Number: 6109-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR