5-Bromo-3-iodo-2-methoxybenzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

054250-500MG 054250-1G 054250-5G
101906-762EA 747.69 USD
101906-762 101906-764 101906-766
5-Bromo-3-iodo-2-methoxybenzenecarboxylic acid ≥95%
5-Bromo-3-iodo-2-methoxybenzenecarboxylic acid

Hazard: Irritant

Formula: C₈H₆BrIO₃
MDL Number: MFCD14581641
CAS Number: 1198475-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR