N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]acetamide ≥95%

Supplier: Matrix Scientific

054247-500MG 054247-1G
101906-754EA 375.38 USD
101906-754 101906-756
N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]acetamide ≥95%
N-[4-Bromo-2-nitro-5-(trifluoromethyl)phenyl]acetamide

Hazard: Irritant

Formula: C₉H₆BrF₃N₂O₃
MDL Number: MFCD09263995
CAS Number: 157554-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR