2-Iodo-6-nitro-4-(trifluoromethyl)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3-iodo-5-nitrobenzotrifluoride

054241-500MG 054241-1G 054241-5G
101906-728EA 375.38 USD
101906-728 101906-730 101906-732
2-Iodo-6-nitro-4-(trifluoromethyl)aniline ≥95%
2-Iodo-6-nitro-4-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₇H₄F₃IN₂O₂
MDL Number: MFCD14560546
CAS Number: 543740-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR