Methyl-5-bromo-3-iodo-2-methoxybenzenecarboxylate ≥95%
Supplier: Matrix Scientific
054239-500MG
054239-1G
054239-5G
101906-718EA
318.46
USD
101906-718
101906-720
101906-722
Methyl-5-bromo-3-iodo-2-methoxybenzenecarboxylate ≥95%
Methyl-5-bromo-3-iodo-2-methoxybenzenecarboxylate
Hazard: Irritant
Formula:
C₉H₈BrIO₃ |
MDL Number:
MFCD14581636 CAS Number: 1155261-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...