Methyl-5-bromo-2-hydroxy-3-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-bromo-2-hydroxy-3-nitrobenzoate

054237-500MG 054237-1G 054237-5G
101906-708EA 375.38 USD
101906-708 101906-710 101906-712
Methyl-5-bromo-2-hydroxy-3-nitrobenzenecarboxylate ≥95%
Methyl-5-bromo-2-hydroxy-3-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₈H₆BrNO₅
MW: 276.05 g/mol
MDL Number: MFCD00458681
CAS Number: 91983-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR