(4,6-Dichloro-3-pyridinyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloro-5-(hydroxymethyl)pyridine, (4,6-Dichloropyridin-3-yl)methanol

054236-500MG 054236-1G
101906-704EA 747.69 USD
101906-704 101906-706
(4,6-Dichloro-3-pyridinyl)methanol ≥95%
(4,6-Dichloro-3-pyridinyl)methanol

Hazard: Irritant

Formula: C₆H₅Cl₂NO
Melting Pt: 82…83 °C
MDL Number: MFCD04114262
CAS Number: 73998-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR