4-Iodo-3-methoxybenzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Iodo-3-methoxybenzoic acid

054235-500MG 054235-1G
101906-700EA 284.62 USD
101906-700 101906-702
4-Iodo-3-methoxybenzenecarboxylic acid ≥95%
4-Iodo-3-methoxybenzenecarboxylic acid

Hazard: Irritant

Formula: C₈H₇IO₃
MW: 278.05 g/mol
MDL Number: MFCD09753732
CAS Number: 282087-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR