(2,3-Dichloro-4-pyridinyl)methanol ≥95%

Supplier: Matrix Scientific

054234-500MG 054234-1G
101906-696EA 747.69 USD
101906-696 101906-698
(2,3-Dichloro-4-pyridinyl)methanol ≥95%
(2,3-Dichloro-4-pyridinyl)methanol

Hazard: Irritant

Formula: C₆H₅Cl₂NO
MDL Number: MFCD14581635
CAS Number: 329794-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR