Ethyl-4-chloro-1-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-chloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

054233-500MG 054233-1G
101906-692EA 423.08 USD
101906-692 101906-694
Ethyl-4-chloro-1-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate ≥95%
Ethyl-4-chloro-1-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₉H₁₀ClNO₃
MW: 215.64 g/mol
MDL Number: MFCD08234670
CAS Number: 821791-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR