4-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidine ≥95%

Supplier: Matrix Scientific

054232-500MG 054232-1G
101906-688EA 921.54 USD
101906-688 101906-690
4-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidine ≥95%
4-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidine

Hazard: Irritant/Refrigerate

Formula: C₇H₇ClN₄
MDL Number: MFCD09901801
CAS Number: 87412-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR