1,3-Dimethyl-1,5-dihydro-4H-pyrazolo-[3,4-d]pyrimidin-4-one ≥95%

Supplier: Matrix Scientific

054229-500MG 054229-1G
101906-680EA 556.92 USD
101906-680 101906-682
1,3-Dimethyl-1,5-dihydro-4H-pyrazolo-[3,4-d]pyrimidin-4-one ≥95%
1,3-Dimethyl-1,5-dihydro-4H-pyrazolo-[3,4-d]pyrimidin-4-one

Hazard: Irritant

Formula: C₇H₈N₄O
Melting Pt: 271…273 °C
MDL Number: MFCD09901802
CAS Number: 87412-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR