2-Fluoro-5-nitrobenzylalcohol ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Fluoro-5-nitrophenyl)methanol

054228-500MG 054228-1G 054228-5G
101906-674EA 181.54 USD
101906-674 101906-676 101906-678
2-Fluoro-5-nitrobenzylalcohol ≥95%
2-Fluoro-5-nitrobenzylalkohol

Hazard: Irritant

Formula: C₇H₆FNO₃
MW: 171.1300 g/mol
MDL Number: MFCD02683502
CAS Number: 63878-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR