7-Amino-2-methylpyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

054227-500MG 054227-1G 054227-5G
101906-668EA 747.69 USD
101906-668 101906-670 101906-672
7-Amino-2-methylpyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%
7-Amino-2-methylpyrazolo[1,5-a]pyrimidine-3-carbonitrile

Hazard: Irritant

Formula: C₈H₇N₅
MDL Number: MFCD14581632
CAS Number: 1203898-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR