7-Hydroxy-2-methyl-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

054225-500MG 054225-1G
101906-658EA 556.92 USD
101906-658 101906-660
7-Hydroxy-2-methyl-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%
7-Hydroxy-2-methyl-5-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carbonitrile

Hazard: Irritant

Formula: C₉H₅F₃N₄O
MDL Number: MFCD14581631
CAS Number: 1198475-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR