2,3-Dichloro-4-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

054224-500MG 054224-2.5G
101906-654EA 392.31 USD
101906-654 102551-408
2,3-Dichloro-4-(trifluoromethyl)pyridine ≥95%
2,3-Dichloro-4-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₂Cl₂F₃N
CAS Number: 89719-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR