5-(4-Fluorophenyl)-3-nitro-2-pyridinylamine ≥95%

Supplier: Matrix Scientific

054223-500MG 054223-1G
101906-650EA 278.46 USD
101906-650 101906-652
5-(4-Fluorophenyl)-3-nitro-2-pyridinylamine ≥95%
5-(4-Fluorophenyl)-3-nitro-2-pyridinylamine

Hazard: Irritant

Formula: C₁₁H₈FN₃O₂
MDL Number: MFCD14581630
CAS Number: 640271-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR