2,2,2-Trifluoro-N-(2-methoxyphenethyl)acetamide ≥95%

Supplier: Matrix Scientific

054219-500MG 054219-1G 054219-5G
101906-632EA 278.46 USD
101906-632 101906-634 101906-636
2,2,2-Trifluoro-N-(2-methoxyphenethyl)acetamide ≥95%
2,2,2-Trifluoro-N-(2-methoxyphenethyl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₂F₃NO₂
MDL Number: MFCD00971680
CAS Number: 304458-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR