5-(Benzyloxy)-3-bromo-2-(hydroxymethyl)-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

054216-500MG 054216-1G
101906-618EA 747.69 USD
101906-618 101906-620
5-(Benzyloxy)-3-bromo-2-(hydroxymethyl)-4(1H)-pyridinone ≥95%
5-(Benzyloxy)-3-bromo-2-(hydroxymethyl)-4(1H)-pyridinone

Hazard: Irritant

Formula: C₁₃H₁₂BrNO₃
MDL Number: MFCD14581626
CAS Number: 132038-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR