[5-(Benzyloxy)-6-bromo-4-iodo-2-pyridinyl]methanol ≥95%
Supplier: Matrix Scientific
054215-500MG
054215-1G
101906-614EA
921.54
USD
101906-614
101906-616
[5-(Benzyloxy)-6-bromo-4-iodo-2-pyridinyl]methanol ≥95%
[5-(Benzyloxy)-6-bromo-4-iodo-2-pyridinyl]methanol
Hazard: Irritant
Formula:
C₁₃H₁₁BrINO₂ |
MDL Number:
MFCD14581625 CAS Number: 1198569-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...