Methyl-3,6-dibromo-2-methoxy-1-naphthoate ≥95%

Supplier: Matrix Scientific

054214-500MG 054214-1G 054214-5G
101906-608EA 375.38 USD
101906-608 101906-610 101906-612
Methyl-3,6-dibromo-2-methoxy-1-naphthoate ≥95%
Methyl-3,6-dibromo-2-methoxy-1-naphthoate

Hazard: Irritant

Formula: C₁₃H₁₀Br₂O₃
MDL Number: MFCD14581624
CAS Number: 1198475-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR