(6-Bromo-5-methoxy-2-pyridinyl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (6-Bromo-5-methoxypyridin-2-yl)methanol

054213-500MG 054213-1G
101906-604EA 384.62 USD
101906-604 101906-606
(6-Bromo-5-methoxy-2-pyridinyl)methanol ≥95%
(6-Bromo-5-methoxy-2-pyridinyl)methanol

Hazard: Irritant

Formula: C₇H₈BrNO₂
MW: 218.06 g/mol
MDL Number: MFCD09702159
CAS Number: 905562-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR