Methyl-5-bromo-2-hydroxy-3-iodobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

054212-500MG 054212-1G 054212-5G
101906-598EA 375.38 USD
101906-598 101906-600 101906-602
Methyl-5-bromo-2-hydroxy-3-iodobenzenecarboxylate ≥95%
Methyl-5-bromo-2-hydroxy-3-iodobenzenecarboxylate

Hazard: Irritant

Formula: C₈H₆BrIO₃
Melting Pt: 126…128 °C
MDL Number: MFCD00464941
CAS Number: 18071-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR