Methyl-5-(4-methoxyphenyl)-5-oxopentanoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-(4-methoxyphenyl)-5-oxopentanoate
054207-500MG
054207-1G
101906-582EA
347.69
USD
101906-582
101906-584
Methyl-5-(4-methoxyphenyl)-5-oxopentanoate ≥95%
Methyl-5-(4-methoxyphenyl)-5-oxopentanoate
Hazard: Irritant
Formula:
C₁₃H₁₆O₄ MW: 236.27 g/mol |
MDL Number:
MFCD00229176 CAS Number: 1847-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...