4-Bromo-2-(bromomethyl)-1-iodobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-iodobenzyl bromide

054206-500MG 054206-1G
101906-578EA 280 USD
101906-578 101906-580
4-Bromo-2-(bromomethyl)-1-iodobenzene ≥95%
4-Bromo-2-(bromomethyl)-1-iodobenzene

Hazard: Irritant

Formula: C₇H₅Br₂I
MW: 375.84 g/mol
MDL Number: MFCD12025360
CAS Number: 495414-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR