5-Amino-1,3-dimethyl-1H-pyrazole-4-carbonitrile ≥95%

Supplier: Matrix Scientific

054202-500MG 054202-1G 054202-5G
101906-562EA 123.08 USD
101906-562 101906-564 101906-566
5-Amino-1,3-dimethyl-1H-pyrazole-4-carbonitrile ≥95%
5-Amino-1,3-dimethyl-1H-pyrazole-4-carbonitrile

Hazard: Irritant

Formula: C₆H₈N₄
MDL Number: MFCD00128277
CAS Number: 54820-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR