3-(tert-Butyl)-6-ethyl-9-methyl-7-oxo-3,9-diazabicyclo[3.3.1]nonane-3,6-dicarboxylate ≥95%

Supplier: Matrix Scientific

054201-500MG 054201-1G 054201-5G
101906-556EA 1109.23 USD
101906-556 101906-558 101906-560
3-(tert-Butyl)-6-ethyl-9-methyl-7-oxo-3,9-diazabicyclo[3.3.1]nonane-3,6-dicarboxylate ≥95%
3-(tert-Butyl)-6-ethyl-9-methyl-7-oxo-3,9-diazabicyclo[3.3.1]nonane-3,6-dicarboxylate

Hazard: Irritant

Formula: C₁₆H₂₆N₂O₅
MDL Number: MFCD12922684
CAS Number: 625434-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR