Methyl-2-hydroxy-5-[2-(trimethylsilyl)ethynyl]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

054200-500MG 054200-1G 054200-5G
101906-550EA 273.85 USD
101906-550 101906-552 101906-554
Methyl-2-hydroxy-5-[2-(trimethylsilyl)ethynyl]benzenecarboxylate ≥95%
Methyl-2-hydroxy-5-[2-(trimethylsilyl)ethynyl]benzenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₆O₃Si
MDL Number: MFCD12922681
CAS Number: 119754-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR