Methyl-6-bromo-4-chloroquinoline-8-carboxylate ≥95%
Supplier: Matrix Scientific
054198-500MG
054198-1G
101906-544EA
556.92
USD
101906-544
101906-546
Methyl-6-bromo-4-chloroquinoline-8-carboxylate ≥95%
Methyl 6-bromo-4-chloroquinoline-8-carboxylate
Hazard: Irritant
Formula:
C₁₁H₇BrClNO₂ |
MDL Number:
MFCD14581623 CAS Number: 1198569-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...