6-Bromo-4-chloro-8-nitroquinoline ≥95%

Supplier: Matrix Scientific

054197-500MG 054197-1G
101906-540EA 556.92 USD
101906-540 101906-542
6-Bromo-4-chloro-8-nitroquinoline ≥95%
6-Bromo-4-chloro-8-nitroquinoline

Hazard: Irritant

Formula: C₉H₄BrClN₂O₂
MDL Number: MFCD14581622
CAS Number: 1198475-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR