2,3,6-Tribromopyridine ≥98%

Supplier: Matrix Scientific

054188-5G 054188-25G
101906-506EA 315.38 USD
101906-506 101906-508
2,3,6-Tribromopyridine ≥98%
2,3,6-Tribromopyridine

Hazard: Irritant

Formula: C₅H₂Br₃N
MDL Number: MFCD00234000
CAS Number: 2402-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR