1-(Furo[3,2-b]pyridin-6-yl)ethanone

Supplier: Matrix Scientific

054182-100MG 054182-500MG
101906-486EA 315.38 USD
101906-486 101906-488
1-(Furo[3,2-b]pyridin-6-yl)ethanone
1-(Furo[3,2-b]pyridin-6-yl)ethanone

Hazard: Irritant

Formula: C₉H₇NO₂
MDL Number: MFCD13563103
CAS Number: 1203499-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR