tert-Butyl (2-(trimethylsilyl)furo[3,2-b]pyridin-6-yl)methylcarbamate

Supplier: Matrix Scientific

054180-100MG 054180-500MG
101906-478EA 364.62 USD
101906-478 101906-480
tert-Butyl (2-(trimethylsilyl)furo[3,2-b]pyridin-6-yl)methylcarbamate
tert-Butyl (2-(trimethylsilyl)furo[3,2-b]pyridin-6-yl)methylcarbamate

Hazard: Irritant

Formula: C₁₆H₂₄N₂O₃Si
MDL Number: MFCD13176661
CAS Number: 1188996-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR