(5-Bromofuro[2,3-b]pyridin-2-yl)methanol

Supplier: Matrix Scientific

054177-100MG 054177-500MG
101906-466EA 364.62 USD
101906-466 101906-468
(5-Bromofuro[2,3-b]pyridin-2-yl)methanol
(5-Bromofuro[2,3-b]pyridin-2-yl)methanol

Hazard: Irritant

Formula: C₈H₆BrNO₂
MDL Number: MFCD13176658
CAS Number: 1188990-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR