2-(Trimethylsilyl)furo[3,2-b]pyridine-6-carbonitrile

Supplier: Matrix Scientific

054175-100MG 054175-500MG
101906-458EA 315.38 USD
101906-458 101906-460
2-(Trimethylsilyl)furo[3,2-b]pyridine-6-carbonitrile
2-(Trimethylsilyl)furo[3,2-b]pyridine-6-carbonitrile

Hazard: Irritant

Formula: C₁₁H₁₂N₂OSi
MDL Number: MFCD13176656
CAS Number: 1188988-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR